Assay Details
Target Gene Details
Entrez Gene ID: | 227700 |
Gene Name: | SH3-domain GRB2-like endophilin B2 |
Gene Aliases: |
SPAS-1 |
Location: |
Chr.2:30344777-30359316 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh3glb2 | NM_001289709.1 | NP_001276638.1 | ||
NM_001289710.1 | NP_001276639.1 | |||
NM_139302.2 | NP_647463.1 | |||
AF426314.1 | AAL24820.1 | |||
AK129455.1 | BAC98265.2 | |||
AK152409.1 | BAE31195.1 | |||
AK152502.1 | ||||
AK155248.1 | BAE33146.1 | |||
AK167762.1 | BAE39795.1 | |||
BC024477.1 | AAH24477.1 | |||
BC028859.1 | AAH28859.1 | |||
EF676083.1 |