Assay Details
Target Gene Details
Entrez Gene ID: | 227700 |
Gene Name: | SH3-domain GRB2-like endophilin B2 |
Gene Aliases: |
SPAS-1 |
Location: |
Chr.2:30344777-30359316 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh3glb2 | NM_001289709.1 | 11 | 1127 | NP_001276638.1 |
NM_001289710.1 | 9 | 1049 | NP_001276639.1 | |
NM_139302.2 | 10 | 1112 | NP_647463.1 | |
AF426314.1 | 10 | 1009 | AAL24820.1 | |
AK129455.1 | 9 | 2490 | BAC98265.2 | |
AK152409.1 | 10 | 1042 | BAE31195.1 | |
AK152502.1 | 9 | 1000 | ||
AK155248.1 | 11 | 1052 | BAE33146.1 | |
AK167762.1 | 10 | 1085 | BAE39795.1 | |
AK207575.1 | ||||
BC024477.1 | 10 | 1027 | AAH24477.1 | |
BC028859.1 | 8 | 1451 | AAH28859.1 | |
EF676083.1 | 10 | 1088 |