Assay Details
Target Gene Details
Entrez Gene ID: | 56508 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 4 |
Gene Aliases: |
1300003D15Rik, 5730402K07Rik, 6330581N18Rik, EPAC 2, Epac-2, Epac2, mKIAA4040 |
Location: |
Chr.2:71981215-72257474 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rapgef4 | NM_001204165.1 | 4 | 405 | NP_001191094.1 |
NM_001204167.1 | 4 | 405 | NP_001191096.1 | |
NM_019688.2 | 4 | 405 | NP_062662.1 | |
AB021132.1 | 4 | 380 | BAB18976.1 | |
AF115480.1 | 4 | 333 | AAD09132.1 | |
AK017479.1 | 4 | 362 | BAB30763.1 | |
AK220281.1 | 4 | 405 | BAD90206.1 | |
BC139822.1 | 4 | 337 |
More Information
Set Membership: |
Intragenic Exonic |