Assay Details
Target Gene Details
Entrez Gene ID: | 18830 |
Gene Name: | phospholipid transfer protein |
Gene Aliases: |
Bpife, OD107 |
Location: |
Chr.2:164839518-164857708 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pltp | NM_011125.2 | NP_035255.1 | ||
AK148330.1 | BAE28485.1 | |||
AK149730.1 | BAE29052.1 | |||
AK190857.1 | ||||
BC003782.1 | AAH03782.1 | |||
CT010317.1 | CAJ18525.1 | |||
U28960.1 | AAA87943.1 | |||
U37226.1 | AAA80542.1 |