Assay Details
Target Gene Details
Entrez Gene ID: | 18798 |
Gene Name: | phospholipase C, beta 4 |
Gene Aliases: |
A930039J07Rik, AI854601, C230058B11Rik |
Location: |
Chr.2:135658437-136014602 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcb4 | NM_013829.2 | NP_038857.1 | ||
AF022804.1 | AAD01752.1 | |||
AF332071.1 | AAK56099.1 | |||
AF332072.1 | AAK56100.1 | |||
AK148001.1 | ||||
BC061201.1 | AAH61201.1 | |||
BC089521.1 | AAH89521.1 | |||
BC129883.1 |
More Information
Set Membership: |
Intragenic Non-exonic |