Assay Details
Target Gene Details
Entrez Gene ID: | 72112 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 14D |
Gene Aliases: |
2010107K19Rik, GBPI-1, Ppp1r14 |
Location: |
Chr.2:119218119-119230067 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r14d | NM_001290796.1 | NP_001277725.1 | ||
NM_028104.4 | NP_082380.1 | |||
AF408400.2 | AAK97532.1 | |||
AK008348.1 | BAB25620.1 | |||
AY179331.1 | AAO47399.1 | |||
BC024547.1 | AAH24547.1 |
More Information
Set Membership: |
Intragenic Non-exonic |