Assay Details
Target Gene Details
Entrez Gene ID: | 228005 |
Gene Name: | peptidyl-prolyl isomerase G (cyclophilin G) |
Gene Aliases: |
AU019516, AU022200, B230312B02Rik, CYP, SRCyp |
Location: |
Chr.2:69722617-69754059 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppig | NM_001081086.1 | NP_001074555.1 | ||
AK088007.1 | ||||
AK134121.1 | BAE22023.1 | |||
AK135178.1 | ||||
BC016126.1 | ||||
BC027420.1 | ||||
BC048905.1 | AAH48905.1 | |||
BC094926.1 | AAH94926.1 | |||
BC150694.1 |
More Information
Set Membership: |
Intragenic Non-exonic |