Assay Details
Target Gene Details
Entrez Gene ID: | 228829 |
Gene Name: | PHD finger protein 20 |
Gene Aliases: |
6820402O20Rik |
Location: |
Chr.2:156196647-156309953 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phf20 | NM_172674.2 | NP_766262.2 | ||
AK033017.1 | BAC28129.1 | |||
BC011337.1 | AAH11337.1 | |||
BC060121.1 | AAH60121.1 |