Assay Details
Target Gene Details
Entrez Gene ID: | 18019 |
Gene Name: | nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2 |
Gene Aliases: |
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp |
Location: |
Chr.2:168476410-168601657 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfatc2 | NM_001136073.2 | NP_001129545.1 | ||
NM_001291168.1 | NP_001278097.1 | |||
NM_001291169.1 | NP_001278098.1 | |||
NM_001291170.1 | NP_001278099.1 | |||
NM_001291171.1 | NP_001278100.1 | |||
NM_001291173.1 | NP_001278102.1 | |||
NM_001291174.1 | NP_001278103.1 | |||
NM_001291175.1 | NP_001278104.1 | |||
NM_001291176.1 | NP_001278105.1 | |||
NM_001291177.1 | NP_001278106.1 | |||
NM_001291178.1 | NP_001278107.1 | |||
NM_010899.3 | NP_035029.2 | |||
AK161174.1 | BAE36226.1 | |||
EU887581.1 | ||||
EU887582.1 | ||||
EU887583.1 | ||||
EU887584.1 | ||||
EU887585.1 | ||||
EU887586.1 | ||||
EU887587.1 | ||||
EU887588.1 | ||||
EU887589.1 | ||||
EU887590.1 | ||||
EU887591.1 | ||||
EU887592.1 | ||||
U36575.1 | AAC52930.1 | |||
U36576.1 | AAC52931.1 |
More Information
Set Membership: |
Intragenic Non-exonic |