Assay Details
Target Gene Details
Entrez Gene ID: | 29812 |
Gene Name: | N-myc downstream regulated gene 3 |
Gene Aliases: |
4833415O14Rik, AI747193, Ndr3 |
Location: |
Chr.2:156927342-156992111 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndrg3 | NM_013865.2 | NP_038893.1 | ||
NM_180956.1 | NP_851287.1 | |||
AB033922.1 | BAA85883.1 | |||
AK002272.1 | ||||
AK004689.1 | BAB23475.1 | |||
AK036277.1 | 11 | 3445 | BAC29369.1 | |
AK037487.1 | BAC29818.1 | |||
AK154927.1 | BAE32930.1 | |||
AK185967.1 | ||||
AK188494.1 | ||||
BC018504.1 | AAH18504.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |