Assay Details
Target Gene Details
Entrez Gene ID: | 56406 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC-2, ASC2, NRC, Ncoa7, PRIP, RAP250, mKIAA0181 |
Location: |
Chr.2:155390656-155473894 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncoa6 | NM_001242558.1 | NP_001229487.1 | ||
NM_019825.3 | NP_062799.2 | |||
AF216186.1 | AAF35860.1 | |||
AK144338.1 | ||||
AK171864.1 | BAE42705.1 | |||
BC083187.1 | AAH83187.1 | |||
JF707634.1 | ||||
JF707635.1 | ||||
JF707636.1 |
More Information
Set Membership: |
Intragenic Non-exonic |