Assay Details
Target Gene Details
Entrez Gene ID: | 59126 |
Gene Name: | NIMA (never in mitosis gene a)-related expressed kinase 6 |
Gene Aliases: |
1300007C09Rik |
Location: |
Chr.2:38511697-38587490 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nek6 | NM_001159631.1 | 3 | 292 | NP_001153103.1 |
NM_021606.3 | 3 | 285 | NP_067619.1 | |
AF218847.1 | 3 | 287 | AAG16653.1 | |
AK004925.1 | 3 | 262 | BAB23676.2 | |
AK011519.1 | 3 | 271 | BAB27673.2 | |
AK018168.1 | 3 | 275 | ||
AK029266.1 | ||||
AK075717.1 | 3 | 259 | BAC35907.1 | |
AK075836.1 | 3 | 271 | BAC35995.1 | |
AK089919.1 | 3 | 246 | BAC40995.1 | |
AK170757.1 | 3 | 292 | BAE42008.1 | |
BC019524.1 | 3 | 354 | AAH19524.1 | |
BY205493.1 |
More Information
Set Membership: |
Intragenic Exonic |