Assay Details
Target Gene Details
Entrez Gene ID: | 75495 |
Gene Name: | MORN repeat containing 5 |
Gene Aliases: |
1700010A17Rik |
Location: |
Chr.2:36049473-36079709 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Morn5 | NM_029309.2 | 5 | 591 | NP_083585.1 |
AK005808.1 | 5 | 579 | BAB24248.1 | |
BC036965.1 | 5 | 597 | AAH36965.1 | |
BC048673.1 | 5 | 591 | AAH48673.1 |