Assay Details
Target Gene Details
Entrez Gene ID: | 59030 |
Gene Name: | McKusick-Kaufman syndrome |
Gene Aliases: |
1300013E18Rik, AI463362, AI957237, Bbs6 |
Location: |
Chr.2:136873780-136891505 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mkks | NM_001141946.1 | NP_001135418.1 | ||
NM_001286981.1 | NP_001273910.1 | |||
NM_001286983.1 | NP_001273912.1 | |||
NM_021527.2 | NP_067502.2 | |||
BY735964.1 | ||||
CJ157733.1 |
Target Gene Details
Entrez Gene ID: | 74243 |
Gene Name: | SLX4 interacting protein |
Gene Aliases: |
2210009G21Rik, 2410004I22Rik, C85661, Slxip |
Location: |
Chr.2:136891218-137082176 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slx4ip | NM_028201.2 | NP_082477.1 | ||
AK010392.1 | BAB26906.1 |
More Information
Set Membership: |
Intragenic Non-exonic |