Assay Details
Target Gene Details
Entrez Gene ID: | 228576 |
Gene Name: | mal, T cell differentiation protein-like |
Gene Aliases: |
BC012256, BENE |
Location: |
Chr.2:127704390-127729897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mall | NM_145532.3 | NP_663507.1 | ||
AK078971.1 | BAC37487.1 | |||
AK163867.1 | BAE37520.1 | |||
BC012256.1 | AAH12256.1 |
More Information
Set Membership: |
Intragenic Non-exonic |