Assay Details
Target Gene Details
Entrez Gene ID: | 228576 |
Gene Name: | mal, T cell differentiation protein-like |
Gene Aliases: |
BC012256, BENE |
Location: |
Chr.2:127704390-127729897 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mall | NM_145532.3 | 2 | 273 | NP_663507.1 |
AK078971.1 | 2 | 274 | BAC37487.1 | |
AK163867.1 | 2 | 274 | BAE37520.1 | |
BC012256.1 | 2 | 308 | AAH12256.1 |
More Information
Set Membership: |
Intragenic Exonic |