Assay Details
Target Gene Details
Entrez Gene ID: | 72899 |
Gene Name: | MACRO domain containing 2 |
Gene Aliases: |
1110033L15Rik, 2610107G07Rik, 2900006F19Rik |
Location: |
Chr.2:140395356-142392966 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Macrod2 | NM_001013802.3 | NP_001013824.2 | ||
NM_028387.1 | NP_082663.1 | |||
AK004072.1 | BAB23153.1 | |||
AK011824.1 | ||||
AK013491.1 | ||||
AK029184.1 | BAC26340.1 | |||
AK134694.1 | BAE22244.1 | |||
AK160433.1 | BAE35786.1 | |||
AK165589.1 | BAE38277.1 | |||
BC027552.1 | ||||
BC028443.1 | ||||
BC038317.1 | AAH38317.1 |