Assay Details
Target Gene Details
Entrez Gene ID: | 72899 |
Gene Name: | MACRO domain containing 2 |
Gene Aliases: |
1110033L15Rik, 2610107G07Rik, 2900006F19Rik |
Location: |
Chr.2:140395356-142392966 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Macrod2 | NM_001013802.3 | NP_001013824.2 | ||
AK134694.1 | BAE22244.1 | |||
BC076574.1 | AAH76574.1 |