Assay Details
Target Gene Details
Entrez Gene ID: | 99010 |
Gene Name: | lysophosphatidylcholine acyltransferase 4 |
Gene Aliases: |
AI505034, Agpat7, Aytl3, LPEAT2 |
Location: |
Chr.2:112239841-112247111 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lpcat4 | NM_207206.2 | 3 | 513 | NP_997089.1 |
BC068131.1 | 3 | 513 | AAH68131.1 | |
BC080829.1 | 3 | 480 | AAH80829.1 |
More Information
Set Membership: |
Intragenic Exonic |