Assay Details
Target Gene Details
Entrez Gene ID: | 170738 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 7 |
Gene Aliases: |
9330137I11Rik, Kv11.3, erg3 |
Location: |
Chr.2:62699587-63184287 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnh7 | NM_133207.2 | 2 | 366 | NP_573470.2 |
AJ291608.1 | 2 | 173 | CAC14797.1 | |
AK034003.1 | 2 | 367 | BAC28543.1 | |
AK043768.1 | 2 | 342 | ||
AK052366.1 | 2 | 250 | BAC34958.1 |
More Information
Set Membership: |
Intragenic Exonic |