Assay Details
Target Gene Details
Entrez Gene ID: | 16401 |
Gene Name: | integrin alpha 4 |
Gene Aliases: |
CD49D, Itga4B |
Location: |
Chr.2:79255426-79333114 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga4 | NM_010576.3 | 4 | 725 | NP_034706.3 |
AB850880.1 | 4 | 490 | ||
AF109136.1 | 4 | 561 | AAC95388.1 | |
AK039959.1 | 4 | 579 | ||
AK051676.1 | 4 | 651 | BAC34715.1 | |
AK078842.1 | 4 | 430 | ||
AK089898.1 | 3 | 525 | ||
AK156015.1 | 4 | 621 | BAE33546.1 | |
AK156517.1 | 4 | 495 | BAE33742.1 | |
BC068313.1 | 4 | 505 | AAH68313.1 | |
U97151.1 | 2 | 93 | AAC05709.1 | |
X53176.1 | 4 | 648 | CAA37316.1 | |
X53177.1 | 2 | 252 |
More Information
Set Membership: |
Intragenic Exonic |