Assay Details
Target Gene Details
Entrez Gene ID: | 16401 |
Gene Name: | integrin alpha 4 |
Gene Aliases: |
CD49D, Itga4B |
Location: |
Chr.2:79255426-79333114 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga4 | NM_010576.3 | NP_034706.3 | ||
AB850880.1 | ||||
AF109136.1 | AAC95388.1 | |||
BC068313.1 | AAH68313.1 | |||
X53176.1 | CAA37316.1 |
More Information
Set Membership: |
Intragenic Non-exonic |