Assay Details
Target Gene Details
Entrez Gene ID: | 227835 |
Gene Name: | glycosyltransferase-like domain containing 1 |
Gene Aliases: |
E330008O22Rik |
Location: |
Chr.2:44564412-44927918 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtdc1 | NM_172662.3 | NP_766250.2 | ||
AK028468.1 | BAC25967.1 | |||
AK054272.1 | BAC35713.1 | |||
BC052069.1 | AAH52069.1 |
More Information
Set Membership: |
Intragenic Non-exonic |