Assay Details
Target Gene Details
Entrez Gene ID: | 15112 |
Gene Name: | hydroxyacid oxidase 1, liver |
Gene Aliases: |
GOX, Gox1, Hao-1 |
Location: |
Chr.2:134497361-134554352 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hao1 | NM_010403.2 | 2 | 232 | NP_034533.1 |
AF104312.1 | 2 | 232 | AAD25332.1 | |
AK149565.1 | 2 | 239 | BAE28963.1 | |
BC119535.1 | 2 | 232 | ||
BC119536.1 | 2 | 232 |
More Information
Set Membership: |
Intragenic Exonic |