Assay Details
Target Gene Details
Entrez Gene ID: | 207182 |
Gene Name: | gamma-glutamyltransferase 7 |
Gene Aliases: |
1110017C11Rik, 6330563L03Rik, Ggtl3 |
Location: |
Chr.2:155490379-155514860 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ggt7 | NM_144786.2 | 7 | 933 | NP_659035.2 |
AK032051.1 | 7 | 931 | BAC27672.1 | |
AK131175.1 | 5 | 1395 | BAD21425.1 | |
BC005772.1 | 7 | 914 | AAH05772.1 |
More Information
Set Membership: |
Intragenic Exonic |