Assay Details
Target Gene Details
Entrez Gene ID: | 227960 |
Gene Name: | grancalcin |
Gene Aliases: |
5133401E04Rik, AI573844 |
Location: |
Chr.2:62664327-62694109 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gca | NM_145523.3 | NP_663498.1 | ||
AB088388.1 | BAC07231.1 | |||
AF518325.1 | AAM66720.1 | |||
AK033740.1 | BAC28458.1 | |||
AK046347.1 | BAC32686.1 | |||
AK049317.1 | BAC33679.1 | |||
AK050540.1 | BAC34315.1 | |||
AK078915.1 | BAC37457.1 | |||
BC021450.1 | AAH21450.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |