Assay Details
Target Gene Details
Entrez Gene ID: | 15376 |
Gene Name: | forkhead box A2 |
Gene Aliases: |
HNF3-beta, HNF3beta, Hnf-3b, Hnf3b, Tcf-3b, Tcf3b |
Location: |
Chr.2:148042877-148046969 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxa2 | NM_001291065.1 | 1 | 46 | NP_001277994.1 |
NM_010446.3 | NP_034576.2 | |||
AB231453.1 | ||||
CF948812.1 | ||||
L10409.1 | AAA03161.1 | |||
X74937.1 | CAA52891.1 |
More Information
Set Membership: |
Intragenic Exonic |