Assay Details
Target Gene Details
Entrez Gene ID: | 71436 |
Gene Name: | fibronectin leucine rich transmembrane protein 3 |
Gene Aliases: |
5530600M07Rik, C430047I10Rik, mKIAA1469 |
Location: |
Chr.2:140658198-140671476 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Flrt3 | NM_001172160.1 | 1 | 166 | NP_001165631.1 |
NM_178382.4 | 1 | 166 | NP_848469.1 | |
AK028252.1 | 1 | 160 | BAC25843.1 | |
AK031464.1 | 1 | 160 | BAC27417.1 | |
AK045146.1 | 1 | 140 | ||
AK129369.1 | 1 | 90 | BAC98179.1 | |
AK161100.1 | 1 | 160 | ||
BC052043.1 | 1 | 141 | AAH52043.1 |
Target Gene Details
Entrez Gene ID: | 72899 |
Gene Name: | MACRO domain containing 2 |
Gene Aliases: |
1110033L15Rik, 2610107G07Rik, 2900006F19Rik |
Location: |
Chr.2:140395356-142392966 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Macrod2 | NM_001013802.3 | NP_001013824.2 | ||
AK013491.1 | ||||
AK134694.1 | BAE22244.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |