Assay Details
Target Gene Details
Entrez Gene ID: | 72090 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
- |
Location: |
Chr.2:25080146-25085719 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Entpd8 | NM_028093.1 | NP_082369.1 | ||
AK008590.1 | ||||
AY364442.1 | AAQ84519.1 | |||
BC031143.1 |
More Information
Set Membership: |
Intragenic Non-exonic |