Assay Details
Target Gene Details
Entrez Gene ID: | 13480 |
Gene Name: | dolichol-phosphate (beta-D) mannosyltransferase 1 |
Gene Aliases: |
AI118379, AI194292 |
Location: |
Chr.2:168209048-168230379 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpm1 | NM_010072.3 | NP_034202.1 | ||
AB004789.1 | BAA25759.1 | |||
AI099209.1 | ||||
AK004834.1 | BAB23602.1 | |||
AK005268.1 | BAB23920.1 | |||
AK008548.1 | BAB25735.1 | |||
AK147481.1 | 1 | 2417 | ||
BC061151.1 | AAH61151.1 |
More Information
Set Membership: |
Intragenic Non-exonic |