Assay Details
Target Gene Details
Entrez Gene ID: | 228714 |
Gene Name: | cysteine and glycine-rich protein 2 binding protein |
Gene Aliases: |
2510008M08Rik, ATAC2, AU023459, D2Ertd473e, D2Wsu131e, E430020F17 |
Location: |
Chr.2:144369032-144407675 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csrp2bp | NM_001166640.1 | 3 | 358 | NP_001160112.1 |
NM_181417.3 | 3 | 841 | NP_852082.2 | |
BC031563.1 | 3 | 841 | AAH31563.1 |