Assay Details
Target Gene Details
Entrez Gene ID: | 66971 |
Gene Name: | CDK5 regulatory subunit associated protein 1 |
Gene Aliases: |
2310066P17Rik |
Location: |
Chr.2:154289809-154372850 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdk5rap1 | NM_025876.2 | NP_080152.1 | ||
AK010081.1 | ||||
AK010097.1 | BAB26700.1 | |||
AK088517.1 | BAC40398.1 | |||
BC025132.1 | AAH25132.1 |