Assay Details
Target Gene Details
Entrez Gene ID: | 320664 |
Gene Name: | Cas scaffolding protein family member 4 |
Gene Aliases: |
F730031O20Rik |
Location: |
Chr.2:172393683-172433757 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cass4 | NM_001080820.2 | NP_001074289.1 | ||
NM_001276422.1 | NP_001263351.1 | |||
NM_001276423.1 | NP_001263352.1 | |||
AK036419.1 | ||||
AK080960.1 | ||||
BC117858.1 | ||||
BC117859.1 | ||||
BC129975.1 | ||||
BC129976.1 |