Assay Details
Target Gene Details
Entrez Gene ID: | 407823 |
Gene Name: | bromodomain adjacent to zinc finger domain, 2B |
Gene Aliases: |
5830435C13Rik, BC053917, D2Ertd794e |
Location: |
Chr.2:59899363-60209846 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz2b | NM_001001182.3 | NP_001001182.2 | ||
AK029231.1 | ||||
AK030867.1 | BAC27164.1 | |||
AK134250.1 | ||||
AK173180.1 | BAD32458.1 | |||
BC150814.1 |
More Information
Set Membership: |
Intragenic Non-exonic |