Assay Details
Target Gene Details
Entrez Gene ID: | 12075 |
Gene Name: | beaded filament structural protein 1, in lens-CP94 |
Gene Aliases: |
CP95 |
Location: |
Chr.2:143826528-143915274 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bfsp1 | NM_001291061.1 | 6 | 1086 | NP_001277990.1 |
NM_009751.2 | 6 | 1068 | NP_033881.2 | |
AB003147.1 | 6 | 856 | BAA24139.1 | |
AK053766.1 | 6 | 839 | BAC35514.1 | |
AK143180.1 | 6 | 839 | BAE25294.1 | |
BC057580.1 | 6 | 1068 | AAH57580.1 |