Assay Details
Target Gene Details
Entrez Gene ID: | 228788 |
Gene Name: | cerebral cavernous malformation 2-like |
Gene Aliases: |
- |
Location: |
Chr.2:153065933-153081735 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccm2l | NM_145536.3 | NP_663511.2 | ||
AK029326.1 | 1 | 3699 | ||
BC020535.1 | AAH20535.1 |