Assay Details
Target Gene Details
Entrez Gene ID: | 11909 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP |
Location: |
Chr.2:73816509-73892639 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atf2 | NM_001025093.2 | NP_001020264.1 | ||
NM_001284369.1 | NP_001271298.1 | |||
NM_001284372.1 | NP_001271301.1 | |||
NM_001284373.1 | NP_001271302.1 | |||
NM_001284374.1 | NP_001271303.1 | |||
NM_001284376.1 | NP_001271305.1 | |||
NM_009715.3 | NP_033845.1 | |||
AF483482.1 | AAL90756.1 | |||
AF483483.1 | AAL90757.1 | |||
AK046184.1 | BAC32623.1 | |||
AK051591.1 | BAC34684.1 | |||
AK139515.1 | ||||
AK144977.1 | ||||
AK145887.1 | BAE26726.1 | |||
AY902311.1 | AAX83926.1 | |||
BC042210.1 | AAH42210.1 | |||
BC079883.1 | AAH79883.1 | |||
BC082596.1 | AAH82596.1 | |||
M31629.1 | AAA39780.1 | |||
M77167.1 | AAA40394.1 | |||
U46026.1 | AAB64016.1 |
More Information
Set Membership: |
Intragenic Non-exonic |