Assay Details
Target Gene Details
Entrez Gene ID: | 75894 |
Gene Name: | adenosine deaminase-like |
Gene Aliases: |
4930578F03Rik |
Location: |
Chr.2:121140428-121156680 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adal | NM_001290811.1 | NP_001277740.1 | ||
NM_001290812.1 | NP_001277741.1 | |||
NM_029475.2 | NP_083751.1 | |||
AK016299.1 | BAB30184.1 | |||
AK039677.1 | ||||
AK044025.1 | BAC31744.1 | |||
AK048809.1 | BAC33464.1 | |||
AK049697.1 | BAC33880.1 | |||
BC050879.1 | AAH50879.1 | |||
BC052048.1 | AAH52048.1 |
More Information
Set Membership: |
Intragenic Non-exonic |