Assay Details
Target Gene Details
Entrez Gene ID: | 215456 |
Gene Name: | glycerol-3-phosphate acyltransferase 2, mitochondrial |
Gene Aliases: |
A530057A03Rik, Gm116, xGPAT1 |
Location: |
Chr.2:127410628-127436093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpat2 | NM_001081089.2 | NP_001074558.2 | ||
AB257502.1 | ||||
AK131154.1 | BAD21404.1 | |||
BC113165.1 | ||||
BC113776.1 |
More Information
Set Membership: |
Intragenic Non-exonic |