Assay Details
Target Gene Details
Entrez Gene ID: | 11477 |
Gene Name: | activin A receptor, type 1 |
Gene Aliases: |
ALK2, ActR-I, ActRIA, Acvr, Acvrlk2, Alk-2, Alk8, D330013D15Rik, SKR1, Tsk7L |
Location: |
Chr.2:58446438-58566828 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acvr1 | NM_001110204.1 | NP_001103674.1 | ||
NM_001110205.1 | NP_001103675.1 | |||
NM_007394.3 | NP_031420.2 | |||
AF332087.1 | AAK56115.1 | |||
AF332088.1 | AAK56116.1 | |||
AK053428.1 | ||||
AK150014.1 | BAE29239.1 | |||
AK150075.1 | BAE29286.1 | |||
BC058718.1 | AAH58718.1 | |||
L15436.1 | AAA40495.1 |
More Information
Set Membership: |
Intragenic Non-exonic |