Assay Details
Target Gene Details
Entrez Gene ID: | 76161 |
Gene Name: | lysosomal-associated membrane protein family, member 5 |
Gene Aliases: |
3110035N03Rik, 6330527O06Rik, BAD-LAMP, LAMP-5 |
Location: |
Chr.2:136051959-136069917 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lamp5 | NM_029530.2 | NP_083806.2 | ||
AK014127.1 | BAB29169.1 | |||
AK018222.1 | ||||
AK045946.1 | ||||
BC004791.1 | AAH04791.1 |
More Information
Set Membership: |
Intragenic Non-exonic |