Assay Details
Target Gene Details
Entrez Gene ID: | 629499 |
Gene Name: | maestro heat-like repeat family member 8 |
Gene Aliases: |
4922505G16Rik |
Location: |
Chr.2:157208548-157279549 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mroh8 | NM_001039557.4 | 18 | 2588 | NP_001034646.2 |
AK029507.1 | 18 | 2560 | ||
AK132763.1 | 18 | 2345 | BAE21344.1 | |
BC150937.1 | 18 | 2312 |