Assay Details
Target Gene Details
Entrez Gene ID: | 108989 |
Gene Name: | translocated promoter region, nuclear basket protein |
Gene Aliases: |
2610029M07Rik, C77892 |
Location: |
Chr.1:150392838-150449935 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpr | NM_133780.3 | 15 | 1935 | NP_598541.3 |
AK088078.1 | 14 | 1816 | BAC40132.1 | |
AK132381.1 | 14 | 1837 | BAE21135.1 | |
AK157716.1 | 14 | 1796 | BAE34164.1 | |
AK157974.1 | 14 | 1511 | BAE34294.1 | |
BC021320.1 | 14 | 1705 | ||
BC108349.1 | 14 | 1789 | AAI08350.1 | |
BC141406.1 | 15 | 1800 | ||
BC141410.1 | 15 | 1800 |
More Information
Set Membership: |
Intragenic Exonic |