Assay Details
Target Gene Details
Entrez Gene ID: | 18802 |
Gene Name: | phospholipase C, delta 4 |
Gene Aliases: |
4921507K24Rik |
Location: |
Chr.1:74541972-74565977 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcd4 | NM_001081456.1 | 6 | 785 | NP_001074925.1 |
NM_148937.2 | 6 | 936 | NP_683739.2 | |
AK016945.1 | 6 | 933 | BAB30513.1 | |
AK029561.1 | 6 | 931 | ||
AK039149.1 | 6 | 763 | BAC30256.1 | |
AK140231.1 | 6 | 772 | BAE24292.1 | |
AY033991.1 | 6 | 768 | AAK61537.1 | |
BC066156.1 | 6 | 676 | AAH66156.1 |
More Information
Set Membership: |
Intragenic Exonic |