Assay Details
Target Gene Details
Entrez Gene ID: | 226982 |
Gene Name: | eukaryotic translation initiation factor 5B |
Gene Aliases: |
A030003E17Rik, BC018347, IF2 |
Location: |
Chr.1:37998000-38055579 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif5b | NM_198303.2 | 5 | 1353 | NP_938045.2 |
AK145146.1 | 5 | 1240 | BAE26259.1 | |
AK162176.1 | 4 | 933 | ||
AK168377.1 | 5 | 1234 | ||
BC040746.1 | 5 | 1224 | AAH40746.1 | |
BC060288.1 | 5 | 1193 | AAH60288.1 |
More Information
Set Membership: |
Intragenic Exonic |