Assay Details
Target Gene Details
Entrez Gene ID: | 269180 |
Gene Name: | inositol polyphosphate-4-phosphatase, type I |
Gene Aliases: |
107kDa, 9630012D15, D130048C09Rik, R74740 |
Location: |
Chr.1:37299838-37410740 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inpp4a | NM_001290797.1 | 1 | 206 | NP_001277726.1 |
NM_030266.4 | 1 | 206 | NP_084542.2 | |
AK089671.1 | 1 | 143 | ||
AK149414.1 | 1 | 131 | BAE28860.1 | |
AK162581.1 | 1 | 152 | ||
BY274715.1 | 1 | 149 |