Assay Details
Target Gene Details
Entrez Gene ID: | 208727 |
Gene Name: | histone deacetylase 4 |
Gene Aliases: |
4932408F19Rik, AI047285 |
Location: |
Chr.1:91928779-92180385 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac4 | NM_207225.1 | 11 | 1473 | NP_997108.1 |
AK029933.1 | 8 | 1010 | BAE43272.1 | |
AK155250.1 | 12 | 1659 | BAE33147.1 | |
BC066052.1 | 11 | 1473 | AAH66052.1 |
More Information
Set Membership: |
Intragenic Exonic |