Assay Details
Target Gene Details
Entrez Gene ID: | 226409 |
Gene Name: | zinc finger, RAN-binding domain containing 3 |
Gene Aliases: |
4933425L19Rik, AH2, AI316834, C730006D09 |
Location: |
Chr.1:127954179-128103049 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zranb3 | NM_001285945.1 | 14 | 2122 | NP_001272874.1 |
NM_027678.3 | 14 | 2261 | NP_081954.1 | |
AK050039.1 | 10 | 1542 | BAC34042.1 | |
AK083641.1 | 14 | 2240 | BAC38978.1 | |
BC066035.1 | 14 | 2198 | AAH66035.1 | |
BC117921.1 | 14 | 2076 | ||
BC117922.2 | 14 | 3268 |
More Information
Set Membership: |
Intragenic Exonic |