Assay Details
Target Gene Details
Entrez Gene ID: | 319651 |
Gene Name: | ubiquitin specific peptidase 37 |
Gene Aliases: |
4932415L06Rik, C330008N13Rik |
Location: |
Chr.1:74435510-74544286 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp37 | NM_176972.4 | 11 | 1367 | NP_795946.2 |
AK030013.1 | 11 | 1368 | BAC26734.1 | |
AK035640.1 | 10 | 1351 | BAC29135.1 | |
AK173210.1 | 7 | 843 | BAD32488.1 | |
BC139091.1 | 11 | 1309 |
More Information
Set Membership: |
Intragenic Exonic |