Assay Details
Target Gene Details
Entrez Gene ID: | 108989 |
Gene Name: | translocated promoter region, nuclear basket protein |
Gene Aliases: |
2610029M07Rik, C77892 |
Location: |
Chr.1:150392838-150449935 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpr | NM_133780.3 | 19 | 2618 | NP_598541.3 |
AK132381.1 | 18 | 2520 | BAE21135.1 | |
AK157974.1 | 18 | 2194 | BAE34294.1 | |
BC108349.1 | 18 | 2472 | AAI08350.1 | |
BC141406.1 | 19 | 2483 | ||
BC141410.1 | 19 | 2483 |
More Information
Set Membership: |
Intragenic Exonic |